Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026697.3 | NP_080973.1 | 4 | 536 | ||
GenBank mRNA | AB232609.1 | - | 3 | 167 | ||
AJ968955.1 | - | 4 | 391 | |||
AJ968956.1 | - | 3 | 337 | |||
AJ968957.1 | - | 4 | 391 | |||
AJ968958.1 | - | 3 | 337 | |||
AJ968959.1 | - | 4 | 391 | |||
AJ968960.1 | - | 3 | 337 | |||
AK002704.1 | - | 4 | 408 | |||
AK013410.1 | - | 4 | 389 | |||
AK083451.1 | - | 1 | 1407 | |||
AK133963.1 | - | 4 | 436 | |||
AK135751.1 | - | 4 | 425 | |||
AK160859.1 | - | 4 | 536 | |||
AK165812.1 | - | 4 | 383 | |||
AK168551.1 | - | 4 | 393 | |||
AK168867.1 | - | 4 | 405 | |||
AK169396.1 | - | 4 | 407 | |||
BC009085.1 | - | 4 | 219 | 2649852 | ||
BC025139.1 | - | 4 | 407 | 3980228 | ||
BC056648.1 | - | 4 | 409 | 5034519 |