Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026718.2 | NP_080994.2 | 8 | 961 | ||
GenBank mRNA | AK039432.1 | - | 8 | 1156 | ||
AK156539.1 | - | 8 | 1154 | |||
BC027765.1 | - | 6 | 633 | |||
BC049187.1 | - | 8 | 1140 | |||
BC061692.1 | - | 8 | 961 | 6308996 | ||
BC108413.1 | - | 8 | 1436 | 6487251 |