Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026718.2 | NP_080994.2 | 12 | 1358 | ||
GenBank mRNA | AK039432.1 | - | 12 | 1553 | ||
AK156539.1 | - | 12 | 1551 | |||
AK162340.1 | - | 4 | 379 | |||
BC003286.1 | - | 2 | 85 | |||
BC027765.1 | - | 10 | 1033 | |||
BC049187.1 | - | 12 | 1537 | |||
BC061692.1 | - | 12 | 1358 | 6308996 | ||
BC108413.1 | - | 12 | 1833 | 6487251 |