Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Steap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085409.1 | NP_001078878.1 | 3 | 647 | ||
NM_133186.3 | NP_573449.2 | 2 | 274 | |||
GenBank mRNA | AK003108.1 | - | 250 | |||
AK028961.1 | - | 2 | 250 | |||
AK078769.1 | - | 2 | 668 | |||
AK167028.1 | - | 3 | 647 | |||
AK171237.1 | - | 2 | 279 | |||
AK203257.1 | - | 1 | 181 | |||
AY029586.1 | - | 2 | 262 | |||
AY214462.1 | - | 640 | ||||
BC027251.1 | - | 2 | 257 | |||
BC037435.1 | - | 2 | 275 | 2651584 | ||
BY718114.1 | - | 2 | 254 |