Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005921.1 | NP_005912.1 | 3 | 1302 | ||
XM_017009484.1 | XP_016864973.1 | 3 | 4013 | |||
XM_017009485.1 | XP_016864974.1 | 3 | 741 | |||
GenBank mRNA | AF042838.1 | - | 3 | 744 | ||
AK307828.1 | - | 4 | 522 |