Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023464.2 | NP_075953.1 | 3 | 798 | ||
GenBank mRNA | AB041656.1 | - | 3 | 792 | ||
AK004482.1 | - | 1 | 677 | |||
AK088418.1 | - | 3 | 730 | |||
AK158412.1 | - | 913 | ||||
AK183655.1 | - | 1 | 347 | |||
BC023033.1 | - | 3 | 761 | |||
BC037081.1 | - | 3 | 775 | 5098653 | ||
BC048360.1 | - | 3 | 777 | 5067090 |