Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1110007C09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026738.2 | NP_081014.1 | 2 | 193 | ||
GenBank mRNA | AK003526.1 | - | 2 | 194 | ||
AK154517.1 | - | 2 | 165 | |||
BC023485.1 | - | 3 | 325 | |||
BC047205.1 | - | 2 | 233 | |||
BC147073.1 | - | 2 | 149 | 9056051 | ||
BC147074.1 | - | 2 | 149 | 9056042 |