Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178065.4 | NP_835166.3 | 14 | 2318 | ||
GenBank mRNA | AB093230.1 | - | 13 | 1937 | ||
AK077015.1 | - | 13 | 1850 | |||
AK133332.1 | - | 13 | 1854 | |||
AK147491.1 | - | 14 | 2319 | |||
AK147878.1 | - | 14 | 2282 | |||
AK154529.1 | - | 14 | 2299 | |||
BC006074.1 | - | 5 | 473 | |||
BC049900.1 | - | 8 | 1008 | |||
BC057351.1 | - | 13 | 2146 | |||
BC060658.1 | - | 13 | 1887 | 30355406 | ||
BC079590.1 | - | 14 | 2276 | |||
HM024719.1 | - | 249 |