Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myadml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204820.1 | NP_001191749.1 | 1 | 436 | ||
NM_026751.1 | NP_081027.1 | 1 | 436 | |||
GenBank mRNA | AK003645.1 | - | 1 | 259 | ||
BC091000.1 | - | 1 | 209 | 6756293 | ||
BC109329.1 | - | 1 | 175 | 40051378 | ||
BC125008.1 | - | 1 | 175 | 40051379 |