Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024226.4 | NP_077188.1 | 2, 3 | 537 | ||
NM_194051.3 | NP_918940.1 | 4, 5 | 2824 | |||
NM_194052.3 | NP_918941.1 | 2, 3 | 1013 | |||
NM_194053.3 | NP_918942.1 | 3, 4 | 1070 | |||
NM_194054.3 | NP_918943.1 | 4, 5 | 3431 | |||
GenBank mRNA | AB073672.1 | - | 2, 3 | 1389 | ||
AF326337.1 | - | 2, 3 | 317 | |||
AK034902.1 | - | 2, 3 | 957 | |||
AK211699.1 | - | 2, 3 | 253 | |||
AK212390.1 | - | 2, 3 | 240 | |||
AK220511.1 | - | 4, 5 | 3391 | |||
AY102280.1 | - | 4, 5 | 2824 | |||
AY102281.1 | - | 2, 3 | 970 | |||
AY102282.1 | - | 3, 4 | 1027 | |||
AY102283.1 | - | 2, 3 | 530 | |||
AY102284.1 | - | 4, 5 | 3388 | |||
AY114152.1 | - | 4, 5 | 3389 | |||
AY114153.1 | - | 3, 4 | 844 | |||
BC032192.1 | - | 2, 3 | 1683 | 5323152 | ||
BC032272.1 | - | 2, 3 | 2537 | |||
BC056373.1 | - | 4, 5 | 3259 | |||
BU848611.1 | - | 2, 3 | 376 |