Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040002.1 | NP_001035091.1 | 4 | 771 | ||
NM_004527.3 | NP_004518.1 | 3 | 917 | |||
NM_013999.3 | NP_054705.1 | 2 | 744 | |||
GenBank mRNA | U10492.1 | - | 3 | 917 | ||
U10493.1 | - | 2 | 744 |