Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176833.4 | NP_789803.1 | 5 | 935 | ||
GenBank mRNA | AK016942.1 | - | 5 | 940 | ||
AK154192.1 | - | 5 | 838 | |||
AK164964.1 | - | 5 | 816 | |||
BC042570.1 | - | 4 | 784 | 4241448 |