Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024228.2 | NP_077190.2 | 8 | 807 | ||
GenBank mRNA | AK003726.1 | - | 8 | 809 | ||
BC002172.1 | - | 6 | 726 |