Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023479.2 | NP_075968.2 | 23 | 2279 | ||
GenBank mRNA | AF308696.2 | - | 23 | 2217 | ||
AK003759.1 | - | 5 | 463 | |||
AK004136.1 | - | 11 | 1015 | |||
AK156730.1 | - | 23 | 2280 | |||
BC023134.1 | - | 23 | 2218 | |||
BC048235.1 | - | 23 | 2213 | 6493124 |