Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samm50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178614.4 | NP_848729.1 | 5, 6 | 553 | ||
GenBank mRNA | AK003990.1 | - | 5, 6 | 509 | ||
AK039949.1 | - | 5, 6 | 553 | |||
AK041783.1 | - | 547 | ||||
AK084395.1 | - | 5, 6 | 547 | |||
AK086633.1 | - | 545 | ||||
AK144975.1 | - | 5, 6 | 554 | |||
AK159740.1 | - | 5, 6 | 565 | |||
AK161378.1 | - | 5, 6 | 543 | |||
AK167806.1 | - | 5, 6 | 547 | |||
AK168510.1 | - | 5, 6 | 509 | |||
AK169066.1 | - | 5, 6 | 543 | |||
AK169351.1 | - | 5, 6 | 510 | |||
AK177148.1 | - | 2, 3 | 183 | |||
AK181035.1 | - | 2, 3 | 164 | |||
AK181279.1 | - | 2, 3 | 151 | |||
AK207686.1 | - | 2, 3 | 164 | |||
AK214436.1 | - | 2, 3 | 164 | |||
BC119178.1 | - | 5, 6 | 547 | 8733927 | ||
BC119180.1 | - | 5, 6 | 547 | 8733929 |