Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samm50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178614.4 | NP_848729.1 | 9 | 928 | ||
GenBank mRNA | AK003990.1 | - | 9 | 883 | ||
AK039949.1 | - | 9 | 928 | |||
AK041783.1 | - | 922 | ||||
AK084395.1 | - | 9 | 922 | |||
AK086633.1 | - | 922 | ||||
AK159740.1 | - | 9 | 940 | |||
AK161378.1 | - | 9 | 918 | |||
AK167806.1 | - | 9 | 922 | |||
AK168510.1 | - | 9 | 884 | |||
AK169066.1 | - | 9 | 918 | |||
AK169351.1 | - | 9 | 885 | |||
AK215615.1 | - | 2 | 59 | |||
BC119178.1 | - | 9 | 922 | 8733927 | ||
BC119180.1 | - | 9 | 922 | 8733929 |