Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam172a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163419.1 | NP_001156891.1 | 5 | 566 | ||
NM_138312.1 | NP_612185.1 | 6 | 690 | |||
GenBank mRNA | AF064782.1 | - | 5 | 519 | ||
AK049727.1 | - | 5 | 527 | |||
AK052419.1 | - | 5 | 538 | |||
AK165273.1 | - | 6 | 589 | |||
AK199247.1 | - | 4 | 361 | |||
BC057380.1 | - | 5 | 534 |