Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1110032A03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023483.3 | NP_075972.2 | 4 | 648 | ||
GenBank mRNA | AJ250230.1 | - | 4 | 598 | ||
AK004016.1 | - | 4 | 949 | |||
AK015671.1 | - | 4 | 588 | |||
AK043513.1 | - | 4 | 649 | |||
AK079767.1 | - | 2527 | ||||
AK158460.1 | - | 4 | 637 | |||
AK166202.1 | - | 4 | 613 | |||
BC032012.1 | - | 4 | 622 | |||
CT010205.1 | - | 4 | 395 |