Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1110032A03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023483.3 | NP_075972.2 | 3 | 538 | ||
GenBank mRNA | AJ250230.1 | - | 3 | 488 | ||
AK004016.1 | - | 3 | 839 | |||
AK015671.1 | - | 3 | 478 | |||
AK043513.1 | - | 3 | 539 | |||
AK079767.1 | - | 2417 | ||||
AK158460.1 | - | 3 | 527 | |||
CT010205.1 | - | 3 | 285 |