Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1110032A03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023483.3 | NP_075972.2 | 2 | 431 | ||
GenBank mRNA | AJ250230.1 | - | 2 | 381 | ||
AK004016.1 | - | 2 | 732 | |||
AK015671.1 | - | 2 | 371 | |||
AK043513.1 | - | 2 | 432 | |||
AK079767.1 | - | 1905 | ||||
AK158460.1 | - | 2 | 420 | |||
AK166202.1 | - | 2 | 396 | |||
BC032012.1 | - | 2 | 405 | |||
CT010205.1 | - | 2 | 178 |