Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197987.2 | NP_932104.1 | 21 | 2870 | ||
GenBank mRNA | AB093271.1 | - | 21 | 2841 | ||
AK029570.1 | - | 7 | 1148 | |||
AK036619.1 | - | 5 | 807 | |||
AK086798.1 | - | 8 | 1246 | |||
AK140822.1 | - | 21 | 2871 | |||
BC022117.1 | - | 16 | 2036 | |||
BC024949.1 | - | 2 | 196 | |||
BC058678.1 | - | 21 | 2868 | 6834513 | ||
BC059070.1 | - | 21 | 2836 | 6825999 | ||
BC078416.1 | - | 21 | 2855 |