Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026825.3 | NP_081101.3 | 26 | 2375 | ||
GenBank mRNA | AK043591.1 | - | 11 | 955 | ||
AK049059.1 | - | 8 | 751 | |||
AY437876.1 | - | 26 | 2380 | |||
BC012229.1 | - | 5 | 404 | |||
BC150819.1 | - | 25 | 2289 |