Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080575.2 | NP_542142.1 | 4 | 668 | ||
GenBank mRNA | AB046742.1 | - | 4 | 657 | ||
AK038517.1 | - | 1 | 16 | |||
AK088244.1 | - | 4 | 647 | |||
AK146568.1 | - | 4 | 669 | |||
AK158119.1 | - | 4 | 663 | |||
AK172412.1 | - | 4 | 654 | |||
AK172470.1 | - | 4 | 643 | |||
AK173275.1 | - | 4 | 629 | |||
BC092278.1 | - | 4 | 697 | 3710505 |