Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybphl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026831.1 | NP_081107.1 | 5, 6 | 819 | ||
GenBank mRNA | AK004148.1 | - | 5, 6 | 820 | ||
BC089625.1 | - | 5, 6 | 807 | 6756382 | ||
BC132636.1 | - | 5, 6 | 808 | 40130913 | ||
BC132638.1 | - | 5, 6 | 808 | 40130915 | ||
BY704393.1 | - | 5, 6 | 820 |