Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cgrrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026832.3 | NP_081108.2 | 3 | 382 | ||
GenBank mRNA | AK007402.1 | - | 3 | 398 | ||
AK030403.1 | - | 3 | 383 | |||
AK030755.1 | - | 3 | 384 | |||
AK154797.1 | - | 3 | 458 | |||
BC037677.1 | - | 3 | 398 | 3491666 | ||
BC114536.1 | - | 2 | 220 |