Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005926.2 | NP_005917.2 | 6 | 1059 | ||
GenBank mRNA | AK312867.1 | - | 6 | 1035 | ||
BC023557.2 | - | 6 | 994 | 4139958 | ||
BC050742.1 | - | 6 | 1005 | 6200464 | ||
U04209.1 | - | 6 | 993 |