Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135247.1 | NP_001128719.1 | 7 | 431 | ||
NM_001135248.1 | NP_001128720.1 | 7 | 450 | |||
NM_002403.3 | NP_002394.1 | 7 | 453 | |||
NM_017459.2 | NP_059453.1 | 7 | 434 | |||
XM_011541476.2 | XP_011539778.1 | 7 | 1060 | |||
GenBank mRNA | AK222751.1 | - | 7 | 412 | ||
BC015039.1 | - | 7 | 453 | 3910064 | ||
CR407678.1 | - | 6 | 320 | |||
DN996554.1 | - | 7 | 403 | |||
U19718.1 | - | 6 | 361 |