Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144525.3 | NP_653108.2 | 12 | 1351 | ||
GenBank mRNA | AK034883.1 | - | 12 | 1353 | ||
AK053521.1 | - | 1641 | ||||
AK143078.1 | - | 1353 | ||||
AK165285.1 | - | 11 | 1514 | |||
AK166510.1 | - | 12 | 1346 | |||
AK180903.1 | - | 4 | 259 | |||
AK213740.1 | - | 2 | 90 | |||
BC022142.1 | - | 8 | 628 | 5101205 | ||
BC022602.1 | - | 6 | 497 | 4161204 | ||
BC026651.1 | - | 12 | 1267 | 4987771 | ||
BC027046.1 | - | 6 | 645 | 5362953 | ||
BC029150.1 | - | 12 | 1339 | 4954376 |