Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178615.3 | NP_848730.2 | 2, 3 | 1091 | ||
GenBank mRNA | AJ557514.1 | - | 2, 3 | 1091 | ||
AK004310.1 | - | 2, 3 | 642 | |||
AK030386.1 | - | 2, 3 | 1047 | |||
AK035713.1 | - | 2, 3 | 1038 | |||
AK047390.1 | - | 2, 3 | 1048 | |||
AK080819.1 | - | 2, 3 | 1038 | |||
BC096024.1 | - | 2, 3 | 1038 | 6821755 | ||
BC138890.1 | - | 2, 3 | 914 | 8861912 |