Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nexn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199465.2 | NP_955759.2 | 5 | 427 | ||
GenBank mRNA | AK136097.1 | - | 5 | 553 | ||
AK136102.1 | - | 5 | 355 | |||
AK163414.1 | - | 5 | 427 | |||
AK163848.1 | - | 4 | 341 | |||
BC052878.1 | - | 5 | 374 | 30046316 | ||
HQ179982.1 | - | 5 | 461 |