Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177780.3 | NP_808448.2 | 5 | 513 | ||
GenBank mRNA | AK053909.1 | - | 5 | 384 | ||
AK147975.1 | - | 5 | 374 | |||
BC158046.1 | - | 5 | 373 | 9007351 | ||
BC158135.1 | - | 5 | 373 | 9088200 | ||
BC158138.1 | - | 5 | 373 | 9088202 |