Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017966.2 | NP_001017966.1 | 3 | 512 | ||
GenBank mRNA | AK129015.1 | - | 492 | |||
BC021415.1 | - | 3 | 493 | |||
BY731719.1 | - | 3 | 512 |