Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167767.1 | NP_001161239.1 | 3 | 339 | ||
NM_001167768.1 | NP_001161240.1 | 3 | 328 | |||
NM_172401.4 | NP_765989.3 | 3 | 347 | |||
GenBank mRNA | AI746795.1 | - | 3 | 339 | ||
AK036177.1 | - | 3 | 317 | |||
AK079099.1 | - | 3 | 331 | |||
AK086717.1 | - | 3 | 317 | |||
AK162914.1 | - | 3 | 309 | |||
BB626438.1 | - | 3 | 328 | |||
BC046986.1 | - | 2 | 266 | 6476504 | ||
BC057311.1 | - | 3 | 319 | 6811681 |