Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gkn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026860.1 | NP_081136.1 | 5 | 497 | ||
GenBank mRNA | AK004474.1 | - | 5 | 417 | ||
BC116955.1 | - | 5 | 497 | 40126274 | ||
FN600638.1 | - | 5 | 416 | |||
GU220566.1 | - | 5 | 416 |