Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Disp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278218.1 | NP_001265147.1 | 8 | 1297 | ||
NM_001278219.1 | NP_001265148.1 | 8 | 1257 | |||
NM_001278220.1 | NP_001265149.1 | 7 | 1140 | |||
NM_026866.3 | NP_081142.3 | 9 | 1385 | |||
GenBank mRNA | AK004521.1 | - | 7 | 1115 | ||
AK138276.1 | - | 9 | 1342 | |||
AK170113.1 | - | 8 | 1252 | |||
AY144589.1 | - | 7 | 1095 | |||
AY150577.1 | - | 7 | 1044 | |||
AY150698.1 | - | 9 | 1277 | |||
BC043102.1 | - | 8 | 1258 | 6406376 | ||
BC059225.1 | - | 8 | 1258 | 6406376 |