Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163701.1 | NP_001157173.1 | 3, 4 | 507 | ||
NM_177294.5 | NP_796268.3 | 4, 5 | 584 | |||
GenBank mRNA | AK042776.1 | - | 3, 4 | 485 | ||
AK122505.1 | - | 4, 5 | 551 | |||
AK154007.1 | - | 3, 4 | 490 | |||
AK161487.1 | - | 4, 5 | 567 | |||
AK164658.1 | - | 4, 5 | 567 | |||
AK166232.1 | - | 3, 4 | 490 | |||
AK167589.1 | - | 3, 4 | 490 | |||
AK171233.1 | - | 3, 4 | 463 | |||
AK219445.1 | - | 1, 2 | 133 | |||
BC088899.1 | - | 3, 4 | 470 | |||
BC117973.1 | - | 3, 4 | 476 | 40098643 | ||
BC117974.1 | - | 3, 4 | 476 | 40098644 |