Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166343.1 | NP_001159815.1 | 6 | 995 | ||
NM_002405.3 | NP_002396.2 | 7 | 1090 | |||
NR_029413.1 | 6 | 1041 | ||||
GenBank mRNA | AK292123.1 | - | 7 | 1031 | ||
AK297149.1 | - | 6 | 989 | |||
AK297833.1 | - | 6 | 974 | |||
AK316555.1 | - | 6 | 982 | |||
BC094814.1 | - | 7 | 1051 | 5930073 | ||
CR456518.1 | - | 7 | 883 | |||
U94352.1 | - | 7 | 997 |