Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phkg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026888.3 | NP_081164.2 | 6 | 618 | ||
GenBank mRNA | AA198694.1 | - | 1 | 32 | ||
AK005277.1 | - | 6 | 563 | |||
BC138913.1 | - | 6 | 532 | 8861935 | ||
BC145734.1 | - | 6 | 532 | 40131037 |