Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013256.2 | NP_001013274.2 | 6 | 1190 | ||
NM_001143790.1 | NP_001137262.1 | 6 | 1190 | |||
NM_001281464.1 | NP_001268393.1 | 6 | 1061 | |||
NM_001281465.1 | NP_001268394.1 | 6 | 1174 | |||
GenBank mRNA | AK038651.1 | - | 6 | 977 | ||
AK044055.1 | - | 6 | 969 | |||
AK145919.1 | - | 6 | 1005 | |||
AK165906.1 | - | 6 | 990 | |||
AK211076.1 | - | 1 | 96 | |||
AK211907.1 | - | 1 | 95 | |||
AK212260.1 | - | 1 | 95 | |||
BC060629.1 | - | 2 | 240 | 6847619 | ||
BC066078.1 | - | 6 | 1005 | |||
BC072625.1 | - | 6 | 1018 | |||
BC147495.1 | - | 6 | 988 | 9056325 | ||
BC147497.1 | - | 6 | 988 | 9056354 |