Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026890.2 | NP_081166.1 | 3 | 255 | ||
GenBank mRNA | AK005098.1 | - | 3 | 137 | ||
BC023770.1 | - | 2 | 140 | |||
BC052790.1 | - | 3 | 193 | 1515292 | ||
CN715203.1 | - | 3 | 255 | |||
EF437387.1 | - | 3 | 144 |