Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020469.2 | NP_065202.2 | 7 | 476 | ||
GenBank mRNA | AF134412.1 | - | 7 | 451 | ||
AF134413.1 | - | 7 | 451 | |||
AF134414.1 | - | 7 | 451 | |||
AF134415.1 | - | 7 | 450 | |||
AF134416.1 | - | 7 | 450 | |||
AK302203.1 | - | 6 | 324 | |||
AY579471.1 | - | 1 | 77 | |||
AY727862.1 | - | 2 | 212 | |||
BC069595.1 | - | 7 | 451 | 7262430 | ||
BC069814.1 | - | 7 | 451 | |||
BC111575.1 | - | 7 | 451 | 40069575 | ||
DQ139865.1 | - | 7 | 451 | |||
FR691751.1 | - | 7 | 451 | |||
FR691752.1 | - | 7 | 451 | |||
J05175.1 | - | 7 | 448 | |||
JN791441.1 | - | 7 | 451 | |||
JX398330.1 | - | 7 | 451 | |||
JX398331.1 | - | 7 | 451 | |||
JX398332.1 | - | 7 | 451 | |||
JX398333.1 | - | 7 | 451 | |||
JX519569.1 | - | 7 | 451 | |||
JX519570.1 | - | 7 | 451 | |||
KT282967.1 | - | 7 | 451 |