Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185162.1 | NP_001172091.1 | 3 | 574 | ||
NM_026898.2 | NP_081174.1 | 3 | 806 | |||
GenBank mRNA | AK005105.1 | - | 3 | 773 | ||
BC028850.1 | - | 2 | 538 | 3987279 |