Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021336.4 | NP_067311.4 | 4 | 611 | ||
GenBank mRNA | AF230356.1 | - | 4 | 340 | ||
AK005256.1 | - | 4 | 403 | |||
AK088438.1 | - | 4 | 386 | |||
AK151800.1 | - | 4 | 404 | |||
AK152546.1 | - | 4 | 404 | |||
AK153502.1 | - | 4 | 404 | |||
AK206837.1 | - | 3 | 174 | |||
BC013777.1 | - | 4 | 378 | 2651704 | ||
BC029642.1 | - | 4 | 378 | 3588138 |