Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159714.1 | NP_001153186.1 | 6 | 572 | ||
NM_027328.4 | NP_081604.3 | 6 | 572 | |||
GenBank mRNA | AK005294.1 | - | 6 | 540 | ||
AK033190.1 | - | 6 | 575 | |||
AK033283.1 | - | 6 | 537 | |||
AK044457.1 | - | 6 | 635 | |||
AK051260.1 | - | 6 | 532 | |||
AY040823.1 | - | 6 | 548 | |||
BC018376.1 | - | 6 | 538 | 3673893 | ||
BC057877.1 | - | 5 | 515 | 5347609 | ||
BC061461.1 | - | 6 | 516 | 5717606 | ||
CT010189.1 | - | 5 | 492 |