Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcts1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026902.3 | NP_081178.1 | 2 | 211 | ||
GenBank mRNA | AK005292.1 | - | 2 | 212 | ||
AK087975.1 | - | 2 | 215 | |||
AK138385.1 | - | 2 | 230 | |||
BC010486.1 | - | 2 | 184 | 3483367 | ||
BC037103.1 | - | 2 | 200 |