Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cab39l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026908.3 | NP_081184.3 | 8 | 900 | ||
GenBank mRNA | AK005323.1 | - | 899 | |||
AK013205.1 | - | 7 | 893 | |||
AK030474.1 | - | 8 | 914 | |||
AK053642.1 | - | 8 | 901 | |||
AK076758.1 | - | 8 | 901 | |||
AK076867.1 | - | 8 | 921 | |||
BC016128.1 | - | 6 | 867 | 4911640 | ||
BC016546.1 | - | 8 | 884 | 3595339 |