Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026917.5 | NP_081193.2 | 4 | 747 | ||
GenBank mRNA | AB088044.1 | - | 3 | 513 | ||
AK007355.1 | - | 5 | 1219 | |||
AK018755.1 | - | 4 | 713 | |||
AK046166.1 | - | 4 | 747 | |||
AK080541.1 | - | 4 | 713 | |||
AK169197.1 | - | 694 | ||||
AY166672.1 | - | 3 | 513 | |||
BC005531.1 | - | 5 | 774 | |||
BC018334.1 | - | 3 | 583 | |||
BC025126.1 | - | 4 | 732 | 3669723 | ||
BC052464.1 | - | 4 | 652 | 6834763 |