Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pycr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133705.2 | NP_598466.1 | 4 | 614 | ||
GenBank mRNA | AK007535.1 | - | 4 | 595 | ||
AK150756.1 | - | 2 | 1734 | |||
AK152302.1 | - | 2 | 1734 | |||
AK165618.1 | - | 4 | 589 | |||
AK167622.1 | - | 4 | 570 | |||
AK179744.1 | - | 1 | 70 | |||
AK181506.1 | - | 2 | 170 | |||
AK184426.1 | - | 2 | 170 | |||
AK190918.1 | - | 1 | 70 | |||
BC006882.1 | - | 4 | 590 | 2647612 |