Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1810013L24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081400.3 | NP_001074869.1 | 3 | 890 | ||
GenBank mRNA | AK007485.1 | - | 3 | 890 | ||
BC038008.1 | - | 2 | 327 | |||
BC116778.1 | - | 3 | 873 | 40126097 | ||
BC116780.1 | - | 3 | 873 | 40126099 |