Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnlip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026925.3 | NP_081201.2 | 8 | 815 | ||
GenBank mRNA | AY387690.1 | - | 7 | 787 | ||
BC061061.1 | - | 8 | 815 | 30316303 |